2-Fluoro-1-isopropoxy-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

38615-500MG 38615-1G 38615-5G
77326-788EA 322.39 USD
77326-788 77350-444 77374-620
2-Fluoro-1-isopropoxy-3-(methoxymethyl)benzene ≥97%
2-Fluoro-1-isopropoxy-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₂
MW: 198.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34529946
CAS Number: 2734774-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR