1,3-Dibromo-5-chloro-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

38601-1G 38601-5G 38601-25G
77340-952EA 64.89 USD
77340-952 77363-060 77374-614
1,3-Dibromo-5-chloro-2-ethoxybenzene ≥97%
1,3-Dibromo-5-chloro-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂ClO
Storage Temperature: Ambient
MDL Number: MFCD29043835
CAS Number: 861338-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR