2-Fluoro-4-iodo-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38590-500MG 38590-1G 38590-5G 38590-25G
77326-782EA 95.79 USD
77326-782 77342-610 77363-050 77374-604
2-Fluoro-4-iodo-3-methylbenzoic acid ≥97%
2-Fluoro-4-iodo-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902650
CAS Number: 2385854-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR