2-Fluoro-4-iodo-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38589-500MG 38589-1G 38589-5G 38589-25G
77326-780EA 95.79 USD
77326-780 77342-608 77363-048 77374-602
2-Fluoro-4-iodo-3-methylbenzaldehyde ≥97%
2-Fluoro-4-iodo-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD32902649
CAS Number: 2386329-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR