1,2-Dimethoxy-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

38586-500MG 38586-1G 38586-5G 38586-25G
77326-778EA 194.67 USD
77326-778 77350-442 77363-046 77385-836
1,2-Dimethoxy-4-(trifluoromethyl)benzene ≥97%
1,2-Dimethoxy-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₉F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18395740
CAS Number: 1261835-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR