2-(Benzyloxy)-1-methoxy-4-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

38585-500MG 38585-1G 38585-5G 38585-25G
77326-776EA 161.71 USD
77326-776 77345-868 77363-044 77385-834
2-(Benzyloxy)-1-methoxy-4-(trifluoromethyl)benzene ≥95%
2-(Benzyloxy)-1-methoxy-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃O₂
MW: 282.258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707100
CAS Number: 847148-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR