2-Butoxy-6-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38580-500MG 38580-1G 38580-5G 38580-25G
77326-774EA 176.13 USD
77326-774 77346-554 77363-042 77385-832
2-Butoxy-6-fluorobenzaldehyde ≥97%
2-Butoxy-6-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD32707098
CAS Number: 1694895-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR