1-Bromo-4-(difluoromethoxy)-5-fluoro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

38570-500MG 38570-1G 38570-5G 38570-25G
77326-768EA 119.48 USD
77326-768 77343-596 77363-036 77374-596
1-Bromo-4-(difluoromethoxy)-5-fluoro-2-methylbenzene ≥95%
1-Bromo-4-(difluoromethoxy)-5-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31598113
CAS Number: 2138169-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR