2-(3,5-Dimethylphenoxy)ethanol ≥97%

Supplier: AOBChem USA

Warning

38567-1G 38567-5G 38567-25G
77341-754EA 104.03 USD
77341-754 77363-034 77374-594
2-(3,5-Dimethylphenoxy)ethanol ≥97%
2-(3,5-Dimethylphenoxy)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09754937
CAS Number: 5960-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR