2-(3-(Trifluoromethyl)phenyl)-1,3,4-oxadiazole ≥95%

Supplier: AOBChem USA

Warning

38555-500MG 38555-1G 38555-5G 38555-25G
77326-766EA 147.29 USD
77326-766 77345-258 77363-032 77385-828
2-(3-(Trifluoromethyl)phenyl)-1,3,4-oxadiazole ≥95%
2-(3-(Trifluoromethyl)phenyl)-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₉H₅F₃N₂O
MW: 214.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28680453
CAS Number: 1450828-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR