2,5-Dichloro-3-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

38530-500MG 38530-1G 38530-5G
77336-670EA 391.4 USD
77336-670 77350-436 77374-592
2,5-Dichloro-3-(trifluoromethyl)phenol ≥97%
2,5-Dichloro-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₃O
MW: 230.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786477
CAS Number: 1805480-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR