Methyl 2,5-dichloro-3-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

38529-500MG 38529-1G 38529-5G 38529-25G
77326-764EA 222.48 USD
77326-764 77350-434 77363-030 77385-826
Methyl 2,5-dichloro-3-(trifluoromethyl)benzoate ≥95%
Methyl 2,5-dichloro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₅Cl₂F₃O₂
MW: 273.036 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31698983
CAS Number: 2385325-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR