4,6-Dibromo-3-butoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38521-500MG 38521-1G 38521-5G 38521-25G
77326-762EA 214.24 USD
77326-762 77350-432 77363-028 77385-824
4,6-Dibromo-3-butoxy-2-fluorobenzaldehyde ≥97%
4,6-Dibromo-3-butoxy-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₁Br₂FO₂
MW: 354.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756162
CAS Number: 2764728-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR