4,6-Dibromo-2-fluoro-3-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38517-500MG 38517-1G 38517-5G 38517-25G
77326-758EA 202.91 USD
77326-758 77350-428 77363-024 77385-820
4,6-Dibromo-2-fluoro-3-isopropoxybenzoic acid ≥97%
4,6-Dibromo-2-fluoro-3-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉Br₂FO₃
Storage Temperature: Ambient
MDL Number: MFCD33403781
CAS Number: 2624417-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR