4,6-Dibromo-2-fluoro-3-isopropoxybenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
38515-500MG
38515-1G
38515-5G
38515-25G
77326-756EA
202.91
USD
77326-756
77350-426
77363-022
77385-818
4,6-Dibromo-2-fluoro-3-isopropoxybenzaldehyde ≥97%
4,6-Dibromo-2-fluoro-3-isopropoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₉Br₂FO₂ MW: 339.984 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD33403780 CAS Number: 2624417-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...