1,5-Dibromo-3-ethyl-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

38500-500MG 38500-1G 38500-5G 38500-25G
77326-752EA 130.81 USD
77326-752 77344-330 77363-020 77385-814
1,5-Dibromo-3-ethyl-2-iodobenzene ≥97%
1,5-Dibromo-3-ethyl-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇Br₂I
MW: 389.854 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846039
CAS Number: 1160573-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR