(3-Bromo-5-ethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38498-500MG 38498-1G 38498-5G
77338-742EA 512.94 USD
77338-742 77350-416 77374-582
(3-Bromo-5-ethylphenyl)methanol ≥97%
(3-Bromo-5-ethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23709586
CAS Number: 1427417-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR