Ethyl 3-bromo-5-ethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38493-500MG 38493-1G 38493-25G 38493-5G
77326-746EA 284.28 USD
77326-746 77350-410 77385-808 77374-580
Ethyl 3-bromo-5-ethylbenzoate ≥97%
Ethyl 3-bromo-5-ethylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27948506
CAS Number: 272130-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR