6-Chloro-3-iodo-2-methylimidazo[1,2-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

38483-500MG 38483-1G 38483-5G 38483-25G
77326-742EA 245.14 USD
77326-742 77350-406 77363-014 77385-804
6-Chloro-3-iodo-2-methylimidazo[1,2-a]pyridine ≥97%
6-Chloro-3-iodo-2-methylimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₆ClIN₂
MW: 292.504 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29760925
CAS Number: 1935106-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR