N-(4-Bromophenyl)-3-methoxybenzothioamide ≥95%

Supplier: AOBChem USA

Warning

38480-500MG 38480-1G 38480-5G 38480-25G
77326-740EA 243.08 USD
77326-740 77350-404 77363-012 77385-802
N-(4-Bromophenyl)-3-methoxybenzothioamide ≥95%
N-(4-Bromophenyl)-3-methoxybenzothioamide

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrNOS
Storage Temperature: Ambient
MDL Number: MFCD32632337
CAS Number: 2404733-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR