1,2-Dibromo-5-ethoxy-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

38464-500MG 38464-1G 38464-5G 38464-25G
77326-730EA 146.26 USD
77326-730 77345-156 77363-004 77385-792
1,2-Dibromo-5-ethoxy-3-(trifluoromethyl)benzene ≥97%
1,2-Dibromo-5-ethoxy-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇Br₂F₃O
MW: 347.954 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632335
CAS Number: 2384010-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR