2-(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

38460-1G 38460-5G 38460-25G
77340-664EA 60.77 USD
77340-664 77363-002 77374-574
2-(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4'-Isopropoxy-[1,1'-biphenyl]-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₁H₂₇BO₃
MW: 338.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34705981
CAS Number: 2828440-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR