2-Bromo-5-(methoxymethyl)-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

38458-500MG 38458-1G 38458-5G
77337-494EA 415.09 USD
77337-494 77350-398 77374-572
2-Bromo-5-(methoxymethyl)-4-methylpyridine ≥97%
2-Bromo-5-(methoxymethyl)-4-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632334
CAS Number: 2404734-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR