Ethyl 6-bromo-4-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

38441-500MG 38441-1G 38441-5G 38441-25G
77326-728EA 236.9 USD
77326-728 77350-394 77363-000 77385-790
Ethyl 6-bromo-4-methylnicotinate ≥97%
Ethyl 6-bromo-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD28746000
CAS Number: 1805556-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR