tert-Butyl 6-bromo-4-methylnicotinate ≥97%

Supplier: AOBChem USA

Warning

38439-500MG 38439-1G 38439-25G 38439-5G
77326-726EA 292.52 USD
77326-726 77350-392 77385-788 77374-568
tert-Butyl 6-bromo-4-methylnicotinate ≥97%
tert-Butyl 6-bromo-4-methylnicotinate

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₂
MW: 272.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178510
CAS Number: 2643367-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR