2,4-Dibromo-1-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

38437-500MG 38437-1G 38437-5G
77338-492EA 491.31 USD
77338-492 77350-390 77374-566
2,4-Dibromo-1-isopropylbenzene ≥97%
2,4-Dibromo-1-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Br₂
Storage Temperature: Ambient
MDL Number: MFCD29093966
CAS Number: 58683-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR