Methyl 5-methoxy-6-methylpicolinate ≥97%

Supplier: AOBChem USA

Warning

38432-500MG 38432-1G 38432-5G
77336-354EA 387.28 USD
77336-354 77350-384 77374-562
Methyl 5-methoxy-6-methylpicolinate ≥97%
Methyl 5-methoxy-6-methylpicolinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12032205
CAS Number: 1315361-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR