Methyl 3-fluoro-2-(methoxymethyl)-6-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38429-500MG 38429-1G 38429-5G
77338-712EA 511.91 USD
77338-712 77350-382 77374-558
Methyl 3-fluoro-2-(methoxymethyl)-6-methylbenzoate ≥97%
Methyl 3-fluoro-2-(methoxymethyl)-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403777
CAS Number: 2624417-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR