5-Fluoro-4-methoxy-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

38408-500MG 38408-1G 38408-5G
77338-240EA 470.71 USD
77338-240 77350-370 77374-544
5-Fluoro-4-methoxy-2-methylphenol ≥97%
5-Fluoro-4-methoxy-2-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD22414647
CAS Number: 1824109-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR