2,4-Dibromo-1-(trifluoromethoxy)benzene ≥95%
Supplier: AOBChem USA
|
Warning
|
38398-500MG
38398-1G
38398-5G
38398-25G
77326-702EA
108.15
USD
77326-702
77343-170
77362-986
77374-540
2,4-Dibromo-1-(trifluoromethoxy)benzene ≥95%
2,4-Dibromo-1-(trifluoromethoxy)benzene
Precaución: for Research Use Only
Formula:
C₇H₃Br₂F₃O MW: 319.90 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20233323 CAS Number: 1840-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...