1-Bromo-3-chloro-2-fluoro-5-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

38390-500MG 38390-1G 38390-5G
77337-262EA 410.97 USD
77337-262 77350-366 77374-538
1-Bromo-3-chloro-2-fluoro-5-methoxybenzene ≥95%
1-Bromo-3-chloro-2-fluoro-5-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756138
CAS Number: 1783528-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR