2-(Difluoromethoxy)-1,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

38360-500MG 38360-1G 38360-5G 38360-25G
77326-696EA 153.47 USD
77326-696 77345-504 77362-980 77385-770
2-(Difluoromethoxy)-1,4-difluorobenzene ≥97%
2-(Difluoromethoxy)-1,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₇H₄F₄O
Storage Temperature: Ambient
MDL Number: MFCD22580938
CAS Number: 1404193-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR