tert-Butyl 5-bromo-2-chloroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

38358-500MG 38358-1G 38358-5G 38358-25G
77326-694EA 211.15 USD
77326-694 77350-364 77362-978 77385-768
tert-Butyl 5-bromo-2-chloroisonicotinate ≥97%
tert-Butyl 5-bromo-2-chloroisonicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrClNO₂
MW: 292.557 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33402452
CAS Number: 1609130-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR