(E)-methyl 3-(3,5-dichloro-4-methoxyphenyl)acrylate ≥97%

Supplier: AOBChem USA

Warning

38342-500MG 38342-1G 38342-5G 38342-25G
77326-688EA 260.59 USD
77326-688 77350-356 77362-972 77385-762
(E)-methyl 3-(3,5-dichloro-4-methoxyphenyl)acrylate ≥97%
(E)-methyl 3-(3,5-dichloro-4-methoxyphenyl)acrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀Cl₂O₃
MW: 261.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28251742
CAS Number: 1824875-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR