(4-Ethoxy-2,5-difluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38335-500MG 38335-1G 38335-5G
77339-028EA 545.9 USD
77339-028 77350-352 77374-530
(4-Ethoxy-2,5-difluorophenyl)methanol ≥97%
(4-Ethoxy-2,5-difluorophenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O₂
MW: 188.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503384
CAS Number: 1936170-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR