4-Ethoxy-2,5-difluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

38332-500MG 38332-1G 38332-5G
77338-476EA 490.28 USD
77338-476 77350-348 77374-526
4-Ethoxy-2,5-difluorobenzoic acid ≥95%
4-Ethoxy-2,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503382
CAS Number: 1935223-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR