(3-Methoxy-4-propoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38327-500MG 38327-1G 38327-5G 38327-25G
77326-684EA 180.25 USD
77326-684 77346-752 77362-970 77385-758
(3-Methoxy-4-propoxyphenyl)methanol ≥97%
(3-Methoxy-4-propoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
MW: 196.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06340073
CAS Number: 103859-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR