(5-Bromo-3-chloro-2-methoxypyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

38319-500MG 38319-1G 38319-25G 38319-5G
77326-680EA 293.55 USD
77326-680 77350-346 77385-754 77374-524
(5-Bromo-3-chloro-2-methoxypyridin-4-yl)methanol ≥97%
(5-Bromo-3-chloro-2-methoxypyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₇BrClNO₂
MW: 252.493 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564068
CAS Number: 2734774-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR