1-(Benzyloxy)-2-chloro-4-(methoxymethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

38318-500MG 38318-1G 38318-5G 38318-25G
77326-678EA 144.2 USD
77326-678 77345-068 77362-966 77385-752
1-(Benzyloxy)-2-chloro-4-(methoxymethyl)benzene ≥95%
1-(Benzyloxy)-2-chloro-4-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅ClO₂
MW: 262.731 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632330
CAS Number: 2404733-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR