1-Bromo-2,4-dimethoxy-3-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

38316-500MG 38316-1G 38316-5G
77326-676EA 340.93 USD
77326-676 77350-344 77374-522
1-Bromo-2,4-dimethoxy-3-(methoxymethyl)benzene ≥97%
1-Bromo-2,4-dimethoxy-3-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO₃
MW: 261.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564067
CAS Number: 586392-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR