Ethyl 4-isopropoxy-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

38315-500MG 38315-1G 38315-25G 38315-5G
77326-674EA 304.88 USD
77326-674 77350-342 77385-750 77374-520
Ethyl 4-isopropoxy-2-methylbenzoate ≥97%
Ethyl 4-isopropoxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
Storage Temperature: Ambient
MDL Number: MFCD31771983
CAS Number: 1368033-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR