Methyl 2-fluoro-3-isopropoxy-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

38306-500MG 38306-1G 38306-25G 38306-5G
77326-668EA 284.28 USD
77326-668 77350-338 77385-744 77374-518
Methyl 2-fluoro-3-isopropoxy-5-(trifluoromethyl)benzoate ≥97%
Methyl 2-fluoro-3-isopropoxy-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₄O₃
MW: 280.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902642
CAS Number: 2586126-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR