2-Fluoro-3-isopropoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

38305-500MG 38305-1G 38305-5G 38305-25G
77326-666EA 201.88 USD
77326-666 77350-336 77362-960 77385-742
2-Fluoro-3-isopropoxy-5-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-isopropoxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₄O₃
MW: 266.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707076
CAS Number: 2432848-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR