(2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38304-500MG 38304-1G 38304-5G 38304-25G
77326-664EA 264.71 USD
77326-664 77350-334 77362-958 77385-740
(2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenyl)methanol ≥97%
(2-Fluoro-3-isopropoxy-5-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD33403764
CAS Number: 2624416-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR