1,3-Dibromo-5-fluoro-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

38302-500MG 38302-1G
77340-554EA 1816.92 USD
77340-554 77350-330
1,3-Dibromo-5-fluoro-2-(trifluoromethyl)benzene ≥97%
1,3-Dibromo-5-fluoro-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₂Br₂F₄
MW: 321.892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784688
CAS Number: 1803837-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR