2-(3,5-Difluoro-4-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

38264-1G 38264-5G 38264-25G
77342-118EA 114.33 USD
77342-118 77362-944 77374-508
2-(3,5-Difluoro-4-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3,5-Difluoro-4-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₁BF₂O₃
MW: 298.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403760
CAS Number: 2828445-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR