2-(4-(Benzyloxy)-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

38259-500MG 38259-1G 38259-5G 38259-25G
77326-650EA 89.61 USD
77326-650 77342-344 77362-942 77374-506
2-(4-(Benzyloxy)-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-(Benzyloxy)-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₀H₂₅BO₃
MW: 324.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24039738
CAS Number: 1609259-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR