3-Cyclobutylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

38224-500MG 38224-1G 38224-5G
77339-260EA 571.65 USD
77339-260 77350-320 77374-504
3-Cyclobutylbenzaldehyde ≥95%
3-Cyclobutylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD24396181
CAS Number: 1432621-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR