4-Butoxy-3,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38203-500MG 38203-1G 38203-5G 38203-25G
77326-642EA 202.91 USD
77326-642 77350-314 77362-934 77385-722
4-Butoxy-3,5-dimethylbenzaldehyde ≥97%
4-Butoxy-3,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11188403
CAS Number: 869663-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR