(2,6-Dibromo-3-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

38201-1G 38201-5G 38201-25G
77341-734EA 103 USD
77341-734 77362-932 77374-498
(2,6-Dibromo-3-methoxyphenyl)methanol ≥97%
(2,6-Dibromo-3-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈Br₂O₂
MW: 295.956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403758
CAS Number: 2247439-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR